Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quercetin dihydrate, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate

551600-100MG 551600-100MG
80055-416EA 53.55 USD InStock
80055-416 CA80055-416
Quercetin dihydrate, Sigma-Aldrich®
Quercetin dihydrate
An inhibitor of PI 3-kinase (IC50=3.8µM) and phospholipase A2 (IC50=2µM). Also inhibits mitochondrial ATPase, phosphodiesterases, and protein kinase C. Induces apoptosis in K562, Molt-4, Raji, and MCAS tumor cell lines.


Formula: C₁₅H₁₄O₉
MW: 338.268 g/mol
MDL Number: MFCD00149487
CAS Number: 6151-25-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty