Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium 4-phenylbutyrate, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 4-Phenylbutyric acid sodium salt

567616-100MG 567616-100MG
80054-472EA 120.69 USD InStock
CA80054-472 80054-472
Sodium 4-phenylbutyrate, Sigma-Aldrich®
Sodium 4-phenylbutyrate
A novel antineoplastic agent and transcriptional regulator

Also acts as an inducer of tumor cytostasis and differentiation. Increases fetal hemoglobin production in vitro and in vivo.


Formula: C₁₀H₁₁O₂·Na
MW: 186.2 g/mol
MDL Number: MFCD00800247
CAS Number: 1716-12-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty