Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

C2 Ceramide, Sigma-Aldrich®

Supplier: MilliporeSigma

110145-5MG 110145-5MG
80050-046EA 141.57 USD InStock
CA80050-046 80050-046
C2 Ceramide, Sigma-Aldrich®
C2 Ceramide
Biologically active, cell-permeable, non-physiological ceramide analog

Inhibits cell growth and induces apoptosis in HL-60 cells. Induces intranucleosomal DNA fragmentation. An activator of heterotrimeric protein phosphatase 2A. Stimulates a cytosolic serine/threonine protein phosphatase in T9 cells at concentrations as low as 100nM. Activates a stress activated protein (SAP) kinase in HL-60 human promyelocytic cells.


Formula: C₂₀H₃₉NO₃
MDL Number: MFCD00153903
CAS Number: 3102-57-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty