Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichostatin A (from Streptomyces species), Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: TSA[R-(E,E)]-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide

647925-1MG 647925-1MG
80054-800EA 285.71 USD InStock
CA80054-800 80054-800
Trichostatin A (from Streptomyces species), Sigma-Aldrich®
Trichostatin A
A potent and reversible inhibitor of histone deacetylase

Blocks cell cycle progression at the G1 phase in HeLa cells and induces a 12-fold increase in intracellular levels of gelsolin. Induces reversion of oncogenic ras-transformed NIH 3T3 cells to a normal morphology. Inhibits IL-2 gene expression (IC50=73nM) in Jurkat cells and shows immunosuppressive activity in a mouse model. Down-regulates p57kip2 in Hep 3B cells.


Formula: C₁₇H₂₂N₂O₃
MW: 302.37 g/mol
MDL Number: MFCD03848392
CAS Number: 58880-19-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty