Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxynonenal 10 mg/ml in ethanol ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 4-Hydroxy-2-nonenal4-HNE(±)4-Hydroxynon-2-enal4-hydroxy-2(E)-nonenal4-Hydroxynon-2-enalHNE

M-Clarity™ quality level = MQ100

393204-1MG 393204-1MG
80052-248EA 208.91 USD InStock
CA80052-248 80052-248
4-Hydroxynonenal 10 mg/ml in ethanol ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
4-Hydroxynonenal
A major aldehydic product formed by peroxidation of [omega]-6-unsaturated fatty acids

Regarded as a specific marker of lipid peroxidation. Inhibits proliferation and induces differentiation of HL-60 human leukemic cells. Also induces cell death in murine alveolar macrophages. An inhibitor of state 3 respiration at micromolar levels. Causes a transient increase in cytosolic Ca2+ and irreversibly inhibits Na+, K+-ATPase activity (IC50=120µM).


Formula: C₉H₁₆O₂
MW: 156.22 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
MDL Number: MFCD00211020
CAS Number: 75899-68-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty