Would you like to log in again?
Lysozyme (Muramidase)
Supplier: Pierce Chemical
89833
PI89833EA 254.58 USD InStock
PI89833
Lysozyme (Muramidase)
Enzymes
Thermo Scientific Lysozyme is an enzyme characterized by the ability to break down the bacterial cell wall to improve protein or nucleic acid extraction efficiency.
  • Bacterial cell wall lytic enzyme that improves inclusion body protein purification
  • Compatible with Thermo Scientific Pierce Cell Lysis Reagents
  • White, lyophilized powder
  • Approx. 20000 units/mg dry weight
  • Must be readily soluble at 5 mg/mL in 0.1 M KH2PO4 to give a clear, colorless solution
Lysozyme is an enzyme used to break down bacterial cell walls to improve protein or nucleic acid extraction efficiency. Lysozymes (muramidases) are a family of enzymes with antimicrobial activity characterized by the ability to damage the cell wall of bacteria.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty