Would you like to log in again?
Mouse Recombinant Nitric Oxide Synthase, EMD Millipore
Supplier: EMD Millipore
482777-50U 482777-50U
80053-412EA 569.49 USD InStock
CA80053-412 80053-412
Mouse Recombinant Nitric Oxide Synthase, EMD Millipore
Enzymes
In 25 mM HEPES, containing 8 µM tetrahydro-L-biopterin, 10% glycerol, pH 7.4. Specific activity: >=7 units/mg protein.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty