Would you like to log in again?
Human Catalase (from Erythrocytes)
Supplier: EMD Millipore
219008-1MG 219008-1MG
80050-884EA 221.12 USD InStock
CA80050-884 80050-884
Human Catalase (from Erythrocytes)
Enzymes
In 50 mM Tris, pH 8.0
Specific activity: >=50000 units/mg protein. One unit is defined as the amount of enzyme that will decompose 1.0 µmol of H₂O₂ per minute at 25 °C, pH 7.0. Prepared from blood that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty