Would you like to log in again?
Human Recombinant Caspase-9 (from E. coli)
Supplier: MilliporeSigma
218807-100U 218807-100U
80108-946EA 631.03 USD InStock
CA80108-946 80108-946
Human Recombinant Caspase-9 (from E. coli)
Proteins and Peptides
Member of the interleukin-1[beta] converting enzyme (ICE) family of cysteine proteases that exists in cells as an inactive proenzyme and is activated during the initiation of apoptosis
Functions to trigger the activation of downstream effector caspases and further leads to disassembly of cell structures. Specific activity: >=400units/mg protein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty