Would you like to log in again?
MnTMPyP
Supplier: EMD Millipore
475872-25MG 475872-25MG
80053-228EA 108.87 USD InStock
80053-228 CA80053-228
MnTMPyP
Enzymes
Cell-permeable superoxide dismutase (SOD) mimetic. Catalyzes the dismutation of O2- (rate constant ~4x107 M-1s-1) even in the presence of excess EDTA. Can substitute for SOD in mutants of E. coli lacking this enzyme. MnTMPyP acts in vivo as an NADPH/GSH:O2- oxidoreductase rather than as an SOD mimetic. Also an efficient peroxynitrite reductase when redox coupled with biological antioxidants.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty