Would you like to log in again?
FeTMPyP
Supplier: EMD Millipore
341501-10MG 341501-10MG
80051-608EA 106.51 USD InStock
80051-608 CA80051-608
FeTMPyP
Enzymes
Catalyzes the isomerization of peroxynitrite (ONOO-) to nitrate (NO3-) under physiological conditions and at realistic dose-derived concentrations (Kcat=2.2x106 M-1s-1)
Effectively prevents the oxidation of lipid components by peroxynitrite. Also catalytically decomposes superoxide radicals (O2· -). Irradiation of the aqueous solution at 335 nm releases hydroxyl (OH·) radicals. FeTMPyP can also serve as a potential drug for reactive oxygen species (ROS) detoxification.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty