Would you like to log in again?
Calbiochem® Calpain Inhibitor 3
Supplier: EMD Millipore
Calbiochem®
208722-25MG 208722-25MG
80055-992EA 223.56 USD InStock
80055-992 CA80055-992
Calbiochem® Calpain Inhibitor 3
Proteins and Peptides
Cell-permeable calpain inhibitor
Reduces capsaicin-mediated cell death in cultured dorsal root ganglion. Also blocks the Ca2+-ionophore A23187-induced suppression of neurite outgrowth in isolated hippocampal pyramidal neurons. Exhibits neuroprotective effect in glutamate-induced toxicity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty