Would you like to log in again?
Calbiochem® Granzyme B Inhibitor I, EMD Millipore
Supplier: EMD Millipore
Calbiochem®
368050-1MG 368050-1MG
80051-934EA 111.73 USD InStock
80051-934 CA80051-934
Calbiochem® Granzyme B Inhibitor I, EMD Millipore
Proteins and Peptides
Inhibits the human and murine serine protease granzyme B, found in the cytoplasmic granules of cytotoxic T lymphocytes and natural killer cells and thereby prevents caspase activation
Also inhibits the apoptosis-related DNA fragmentation in lymphocytes by fragmentin 2, a rat lymphocyte granule protease homologous to granzyme B (ID50=300nM).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty