Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Granzyme B Inhibitor II, Calbiochem®

Supplier: MilliporeSigma

368055-1MG 368055-1MG
80051-938EA 109.75 USD InStock
80051-938 CA80051-938
Granzyme B Inhibitor II, Calbiochem®
Granzyme B Inhibitor II
A potent, reversible granzyme B and caspase-8 inhibitor.


Formula: C₂₁H₃₄N₄O₁₀
MW: 502.5 g/mol
CAS Number: 1258003-96-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty