Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DPQ, Calbiochem®

Supplier: MilliporeSigma

300270-1MG
80059-092EA 125.67 USD InStock
80059-092 CA80059-092
DPQ, Calbiochem®
DPQ
Potent and selective inhibitor of poly(ADP-ribose) polymerase (PARP; IC50=40nM).


Formula: C₁₈H₂₆N₂O₂
MW: 302.4
MDL Number: MFCD01866274
CAS Number: 129075-73-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty