Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NS-2028, Calbiochem®

Supplier: MilliporeSigma
Synonyms: 4H-8-Bromo-1,2,4-oxadiazolo(3,4-d)benz(b)(1,4)oxazin-1-one

492030-5MG
80057-176EA 308.69 USD InStock
80057-176 CA80057-176
NS-2028, Calbiochem®
NS-2028
Potent, specific, and irreversible inhibitor of soluble guanylyl cyclase (IC50=30nM for basal and 200nM for NO-stimulated enzyme activity; IC50=17nM for S-nitrosoglutathione-enhanced soluble guanylyl cyclase activity in homogenates of mouse cerebellum).


Formula: C₉H₅BrN₂O₃
MW: 269.1 g/mol
CAS Number: 204326-43-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty