Would you like to log in again?
NG,NG'-Dimethyl-L-arginine dihydrochloride, Calbiochem®
Supplier: MilliporeSigma
311204-25MG 311204-25MG
80058-038EA 109.75 USD InStock
CA80058-038 80058-038
NG,NG'-Dimethyl-L-arginine dihydrochloride, Calbiochem®
NG,NG'-Dimethyl-L-arginine dihydrochloride
Endogenous inhibitor of nitric oxide synthesis in vitro and in vivo. Does not exhibit any significant inhibitory effect on NOS activity.
CAS Number: 65005-57-4
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty