Would you like to log in again?
Diphenyleneiodonium chloride, Calbiochem®
Supplier: MilliporeSigma
Synonyms: Diphenyleneiodonium Chloride
300260-10MG 300260-10MG
80051-406EA 111.73 USD InStock
80051-406 CA80051-406
Diphenyleneiodonium chloride, Calbiochem®
Diphenyleneiodonium chloride
An irreversible endothelial nitric oxide synthase inhibitor. Inhibits iNOS in macrophages (IC50=50nM). Shown to inhibit acetylcholine-induced relaxation of preconstricted rat thoracic aortic rings (IC50=180nM). Blocks K+ and Ca2+ currents in arterial smooth muscle cells. Inhibitor of mitochondrial NADPH-ubiquinone oxidoreductase.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty