Would you like to log in again?
S-Methyl-L-thiocitrulline dihydrochloride, Calbiochem®
Supplier: MilliporeSigma
472804-10MG 472804-10MG
80057-038EA 209.1 USD InStock
80057-038 CA80057-038
S-Methyl-L-thiocitrulline dihydrochloride, Calbiochem®
S-Methyl-L-thiocitrulline dihydrochloride
Exhibits about 17-fold greater selectivity for rat neuronal nitric oxide synthase (IC50=0.3nM) compared to the endothelial enzyme (IC50=5.4µM).
CAS Number: 209589-59-3
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty