Would you like to log in again?
Cyclin-Dependent Protein Kinase Inhibitor Set, Calbiochem®
Supplier: Calbiochem
219428-1SET 219428-1SET
80050-952EA 296.62 USD InStock
CA80050-952 80050-952
Cyclin-Dependent Protein Kinase Inhibitor Set, Calbiochem®
Cell Culture Supplements and Additives
Set contains 5mg of olomoucine (80053-496), 1mg of iso-olomoucine (80053-498), and 1mg of roscovitine (80054-314)
Supplied with an informational sheet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty