Would you like to log in again?
Calbiochem® MG-115
Supplier: EMD Millipore
Calbiochem®
474780-5MG 474780-5MG
80053-192EA 251.41 USD InStock
80053-192 CA80053-192
Calbiochem® MG-115
Proteins and Peptides
Potent reversible proteasome inhibitor (Ki=21nM and 35nM for 20S and 26S proteasome, respectively)
Specifically inhibits the chymotrypsin-like activity of the proteasome and induces apoptosis in Rat-1 and PC12 cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty