Would you like to log in again?
Calbiochem® Proteasome Inhibitor I
Supplier: EMD Millipore
Calbiochem®
539160-1MG 539160-5MG 539160-5MG 539160-1MG
80053-876EA 348.85 USD InStock
80053-876 CA80053-874 CA80053-876 80053-874
Calbiochem® Proteasome Inhibitor I
Proteins and Peptides
Cell-permeable inhibitor of the chymotrypsin-like activity of the multicatalytic proteinase complex (MCP; 20S proteasome) in HT4 cells
Causes the accumulation of ubiquitinated proteins in neuronal cells. Prevents the activation of NF-[kappa]B in response to TNF[alpha] and okadaic acid through inhibition of l[kappa]B-[alpha] degradation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty