Would you like to log in again?
Proteasome Inhibitor III
Supplier: Merck
Calbiochem®
539163-100UG 539163-100UG
80109-738EA 364.03 USD InStock
CA80109-738 80109-738
Proteasome Inhibitor III
Proteins and Peptides
A boronic acid-based, reversible proteasome inhibitor that is structurally similar to MG-132 (80053-194, -196) but displays much higher potency (Ki=0.03nM vs)
4 nM for MG-132).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty