Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Emetine dihydrochloride, Calbiochem®

Supplier: MilliporeSigma

324693-250MG 324693-250MG
80055-236EA 347.83 USD InStock
80055-236 CA80055-236
Emetine dihydrochloride, Calbiochem®
Emetine dihydrochloride
Principal alkaloid of ipecac, isolated from the ground roots of Uragoga ipecacuanha

Irreversibly blocks protein synthesis in eukaryotes by inhibiting the movement of ribosomes along the mRNA. Induces hypotension by blocking adrenoreceptors. Inhibits cytometric anaylsis of DNA replication at the early S-phase.


Formula: C₂₉H₄₀N₂O₄·2HCl
CAS Number: 316-42-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty