Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phorbol-12,13-dibutyrate, Calbiochem®

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


524390-1MG 524390-5MG 524390-5MG 524390-1MG
80055-388EA 143.09 USD InStock
80055-388 80055-390 CA80055-390 CA80055-388
Phorbol-12,13-dibutyrate, Calbiochem®
Phorbol-12,13-dibutyrate
Strong irritant for mouse skin but only moderately active as a tumor promoter

Activates protein kinase C. Stimulates the phosphorylation of Na+,K+- ATPase, thereby inhibiting its activity. Promotes the expression of inducible NOS in cultured hepatocytes. Commonly used in binding studies or in applications requiring high concentrations of phorbol compounds.


Formula: C₂₈H₄₀O₈
MW: 504.6 g/mol
CAS Number: 37558-16-0
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty