Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Glyco-SNAP-2
Supplier: EMD Millipore
362339-5MG 362339-5MG
80051-926EA 204.51 USD InStock
80051-926 CA80051-926
Glyco-SNAP-2
Enzymes
A stable, water-soluble nitric oxide (NO) donor (1/2=27 hours) with properties similar to those of Glyco-SNAP-1 (80051-924). Dissolve in aqueous buffer immediately after use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty