Would you like to log in again?
Illustrated Safety Signs for Work Areas, Nutheme
Supplier: Nutheme
VZC2015
56607-054CS 211.77 USD InStock
56607-054
Illustrated Safety Signs for Work Areas, Nutheme
Signs
Communicate hazards and instructions in laboratories, shops, factories, or wherever safety is a concern.
These illustrated signs provide simple pictorial representations of messages. Color-coding system emphasizes type of hazard. DANGER is red and black; FIRE is red; CAUTION is yellow and black; NOTICE is blue; INSTRUCTIONAL SAFETY is green; RADIATION is purple and black on yellow. Special ANSI Z.535 signs illustrate DANGER and CAUTION according to OSHA regulations. Aluminum signs (silk screened with enamel) are available with adhesive backing, magnetic backing, or no backing. Styrene signs have no backing. Vinyl signs have adhesive backing. All signs measure 12x10". Aluminum and styrene signs have safety-rounded corners with pre-drilled mounting holes.
All signs meet OSHA regulations. Special ANSI signs are made in accordance with ANSI Z.535.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty