Would you like to log in again?
VACUETTE® Urine Collection Accessories, Greiner Bio-One
Supplier: Greiner bio-one
Vacuette®
724310
95057-161CS 301.85 USD InStock
95057-161
VACUETTE® Urine Collection Accessories, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Urine collection accessories include a variety of vessels and kits designed to improve the sample quality and ensure the efficiency of testing procedures.
  • Clean and secure sample transfer
  • Sterile
  • Breakproof and leakproof
Transfer devices enable a clean and secure sample transfer directly into urine tubes. Beakers are sterile, break-proof, and leak-proof. Culture sets include a urine specimen tube treated with boric acid preservative and a urine transfer device (95057-265).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty