Would you like to log in again?
Calbiochem® Glutathione, monoethylester
Supplier: EMD Millipore
Calbiochem®
353905-50MG 353905-50MG
80051-892EA 166.31 USD InStock
80051-892 CA80051-892
Calbiochem® Glutathione, monoethylester
Proteins and Peptides
Cell-permeable derivative of glutathione (GSH) that undergoes hydrolysis by intracellular esterases thereby increasing intracellular GSH concentration in many tissues and cell types. Effective transport of GSH-MEE has been used to protect cells against radiation damage, oxidants, and various toxic compounds including heavy metals. A protective agent against cellular damage such as in cataracts and mitochondrial degeneration.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty