Would you like to log in again?
NG-Hydroxy-L-arginine monoacetate, Calbiochem®
Supplier: MilliporeSigma
399250-5MG 399250-5MG
80052-260EA 184.24 USD InStock
CA80052-260 80052-260
NG-Hydroxy-L-arginine monoacetate, Calbiochem®
NG-Hydroxy-L-arginine monoacetate
A key intermediate in the biosynthesis of nitric oxide by constitutive nitric oxide synthase (cNOS). NOHA can be efficiently oxidized to nitric oxide and citrulline by cytochrome P450. A potent inhibitor of liver and macrophage arginase (Ki=150µM).
CAS Number: 53598-01-9
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty