Would you like to log in again?
Boric Acid Tablets, Chemplex
Supplier: Chemplex
750
15566-084PK 155.54 USD InStock
15566-084
Boric Acid Tablets, Chemplex
Media for Microbiology Additives and Supplements for Microbiology
Very abrasive heat-resistant additive suitable for processing samples that tend to reweld during comminution in metallic grinding vessels.
Frequently used in conjunction with x-ray mix to impart lubricity for “plasticizing” pellets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty