Would you like to log in again?
BioSorb® Biohazard Spill Control Powder, InfeKta
Supplier: InfeKta
BioSorb®
INFBIO-80 INFBIO-16
89030-536CS 437.84 USD InStock
89030-536 89030-534
BioSorb® Biohazard Spill Control Powder, InfeKta
Spill Control Materials
Super absorbent polymers solidify most liquids to gel form
Ideal for use with any liquid specimen (infectious, diagnostic–Category B, or other). Packaged in a screw cap plastic bottle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty