Would you like to log in again?
Accessories for Lounges, ASI
Supplier: Arlington Scientific
20611-821 28206-003 20610-780 20611-805 20611-800 20611-800 20614-816 20611-805 20610-780 20614-816 20611-821
80087-408EA 358.78 USD InStock
80087-408 CA80087-406 CA80087-416 CA80087-404 80087-406 80087-404 CA80087-444 CA80087-410 80087-444 80087-410 CA80087-408
Accessories for Lounges, ASI
Chairs
Accessories for ASI stationary and mobile lounges
The durable, easy-to-clean cover resists mildew, chemicals, and tearing. The warming pad has a large 50.8x91.4cm (20x36") warming area and features convenient, dual-temperature settings for ultimate comfort
Certifications: Warming pad is UL listed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty