Would you like to log in again?
VWR® Biopsy Foam Pads
Supplier: VWR
VWR®
39VWR-100-1
18000-262PK 90.47 USD InStock
18000-262
VWR® Biopsy Foam Pads
Biopsy Pads
Rectangular foam pads are used to hold samples in place and prevent them from being lost during processing.
  • Open-celled foam allows fluids to transfer safely and thoroughly around sample
  • Hold specimen securely
Samples are sandwiched between pads and placed in cassettes with metal or plastic lids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty