Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
KIMAX® Glass Tubing, Medium Wall, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
80400-134
89003-232CS 2153.25 USD InStock
89003-232
KIMAX® Glass Tubing, Medium Wall, Kimble Chase
Tubing Transfer Tubing
KIMAX brand KG-33 borosilicate glass has a low coefficient of expansion and a high resistance to chemical attack
Tubing with glazed ends has a length of 121.9 cm ±0.6 (48" ± ¹/₄). Tubing with cut ends has a length of 122.6 cm ±0.6 (48 ¹/₄" ± ¹/₄).
Certifications: Meets the requirements for Type I Class A, ASTM E438.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty