Would you like to log in again?
TruCLEAN® II Compact Double-Bucket System, Perfex
Supplier: Perfex
TruCLEAN®
30-2B
22939-962EA 1463.02 USD InStock
22939-962
TruCLEAN® II Compact Double-Bucket System, Perfex
Mops
Compact design makes this system ideal for cleaning and disinfecting small areas
System includes two buckets, high-temperature-resistant casters and wheels, stainless steel wringer, mop head and mop frame. The mop head is made from synthetic yarn with looped-end construction. Stainless steel, swivel mop frame features an aluminum handle that adjusts from 91.4–182.9cm (36–72") for hard-to-reach areas. System is autoclavable.
For replacement mop heads and frames, see 22940-020.
A 9L (2.4gal.) bucket, which captures and isolates soiled contaminates, fits inside the larger 30L (7.9gal) bucket that holds the cleaning agent. Versatile design allows use as a single or double-bucket system.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty