Would you like to log in again?
Stainless Steel Round Bucket and Wringer, Micronova
Supplier: Micronova
B-3 W-6
68400-154EA 1554.03 USD InStock
68400-154 68400-162
Stainless Steel Round Bucket and Wringer, Micronova
Mops
Round, stainless steel bucket with 30 L (8 gal) capacity, available with or without heavy-duty 5 cm (2") casters for easy transport. Stainless steel Downpress Wringer for simple string mop wringing.
  • 35 cm (13.75") diameter
  • Stainless steel carrying handle
  • Buckets with casters can be used as a single-bucket mopping system
  • Buckets, wringers, and wringer insert are constructed of stainless steel
  • Designed for use in the cleanroom and controlled environments
  • Entire system is autoclavable
The Micronova stainless steel round buckets are designed for stand alone use or with mop wringers as a compact and maneuverable mopping system. Use with the W-6 Downpress Wringer to wring string mops with an effortless downward press motion.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty