Would you like to log in again?
Corning® Filter Top Tubes, Sterile, Corning
Supplier: Corning
Corning®
430314 430320
28680-120CS 98.26 USD InStock
28680-120 28680-121
Corning® Filter Top Tubes, Sterile, Corning
Filters Bottle Top Filters
Cellulose acetate membrane, PS funnel and PP collection tube.
  • 50 mm diameter membrane
  • Minimises unnecessary transfers by filtering directly into 50 ml centrifuge tube
  • Includes two centrifuge tube stands with each case
  • Each PP centrifuge tube is supplied with an individually wrapped cap for storage
  • Individually packaged, sterile, certified non pyrogenic
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty