Would you like to log in again?
SP Wilmad-LabGlass Pennyhead [ST] Stoppers, Flask Length, SP Industries
Supplier: SP Industries
LG-10320-114 LG-10320-110 LG-10320-112 LG-10320-108 LG-10320-104 LG-10320-100 LG-10320-106 LG-10320-102
14212-540EA 13.41 USD InStock
14212-540 14212-466 14212-344 14211-190 14212-522 14212-426 14210-838 14211-308
SP Wilmad-LabGlass Pennyhead [ST] Stoppers, Flask Length, SP Industries
Stoppers Glass Stoppers
Glass flask length stoppers may be used with volumetric flasks and separatory funnels
Sizes 9 through 27 are solid. Sizes 32 and 38 are hollow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty