Would you like to log in again?
VWR® Pennyhead Stoppers, PTFE
Supplier: Avantor
VWR®
010.3.16 010.3.32 010.3.22 010.3.27
89026-320EA 27.22 USD InStock
89026-320 89026-328 89026-326 89026-324
VWR® Pennyhead Stoppers, PTFE
Stoppers PE/LDPE/HDPE Stoppers
Hollow pennyhead stoppers are for use with standard taper joints.
  • Chemically resistant
  • Non-contaminating
  • PTFE fabrication
Near total chemical resistance and non-contaminating properties make these stoppers ideal for sealing glassware, particularly when preparing solutions in volumetric flasks. Trapezoidal sealing rings provide high vacuum performance and reduce seizure, while the non-stick properties ensure easy removal. Usable to 280°C (536°F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty