Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate ≥97%

Supplier: Ambeed
Warning

A155488-5G
77549-976EA 115 USD InStock
77549-976
Ethyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate ≥97%
Ethyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate
Caution: for Research Use Only


Formula: C₁₀H₉BrN₂O₂
MW: 269.10
MDL Number: MFCD12827886
CAS Number: 1215504-30-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty