Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromomethyl acetate ≥97%

Supplier: Ambeed
Danger

A178876-5G A178876-25G
77550-156EA 154 USD InStock
77550-156 77550-158
Bromomethyl acetate ≥97%
Bromomethyl acetate
Caution: for Research Use Only


Formula: H₃CCOOCH₂Br
MW: 152.98 g/mol
MDL Number: MFCD00000170
CAS Number: 590-97-6
UN: 2924
ADR: 3,8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty