Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrothiophenol ≥95%

Supplier: Ambeed
Danger
Synonyms: 1-Mercapto-4-nitrobenzene4-Nitrobenzene-1-thiolp-Nitrothiophenol4-Nitrobenzenethiol

A200520-25G
77550-282EA 94 USD InStock
77550-282
4-Nitrothiophenol ≥95%
4-Nitrothiophenol
Caution: for Research Use Only


Formula: C₆H₅NO₂S
MDL Number: MFCD00007343
CAS Number: 1849-36-1
UN: 3335
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty