Would you like to log in again?
Monoject™ Oral Medication Syringe, Covidien
Supplier: Covidien
Monoject™
8881907003 8881907102 8881906005 8881901006 8881903010 8881903002
10841-328BX 73.72 USD InStock
10841-328 10841-326 10841-310 15141-092 10841-314 10841-312
Monoject™ Oral Medication Syringe, Covidien
Syringes Syringes, Medical
Made of polypropylene, this clear syringe features permanent, heat-etched dual graduations.
  • Latex free
  • Amber color available for light-sensitive drugs
Syringe does not accept hypodermic needles.
Packaging: Ribbed tip caps are packaged separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty