Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ampicillin solution 10 mg/ml, CellPro™

Supplier: Alkali Scientific
Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

DS0198
104028-972EA 124.6 USD InStock
104028-972
Ampicillin solution 10 mg/ml, CellPro™
Ampicillin sodium salt
Alkali Scientific™ Ampicillin Solution offers researchers a high-quality antibiotic solution for molecular biology applications. Crafted to stringent standards of purity and sterility, this sterile filtered solution ensures reliable and consistent inhibition of bacterial growth in experimental procedures. Ideal for selective bacterial culture and plasmid maintenance, Alkali Scientific™ Ampicillin Solution guarantees optimal results and peace of mind in your laboratory work.

  • For molecular biology
  • Liquid Medium
  • 0.2 μm filtered


Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39
MDL Number: MFCD00064313
CAS Number: 69-52-3
UN: 3248
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty