Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerulenin, Sigma-Aldrich®

Supplier: MilliporeSigma

219557-5MG 219557-5MG
80056-158EA 188 USD InStock
CA80056-158 80056-158
Cerulenin, Sigma-Aldrich®
Cerulenin
An antifungal antibiotic that inhibits sterol and fatty acid biosynthesis

In fatty acid synthesis, reported to bind in equimolar ratio to [beta]-keto-acyl-ACP synthase. In sterol synthesis, inhibits HMG-CoA synthetase activity.


Formula: C₁₂H₁₇NO₃
MW: 223.272 g/mol
MDL Number: MFCD00077686
CAS Number: 17397-89-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty