Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cytochalasin D (from Zygosporium mansoni) ≥98% (by TLC), white solid, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-triene-1,17-dione 21(R)-acetate

250255-1MG 250255-1MG
80055-214EA 234.64 USD InStock
80055-214 CA80055-214
Cytochalasin D (from Zygosporium mansoni) ≥98% (by TLC), white solid, Sigma-Aldrich®
Cytochalasin D
Cell-permeable fungal toxin

About 10-fold more potent than cytochalasin B (80055-212) in inhibiting actin filament function. Does not inhibit monosaccharide transport across cell membranes. Inhibits cytoplasmic division by blocking the formation of contractile microfilaments. Inactivates low conductance K+channels. Also modulates CD4 cross-linking in T lymphocytes and increases intracellular Ca2+ levels. Exhibits antitumor activity.

Soluble in DMSO or EtOH


Formula: C₃₀H₃₇NO₆
MDL Number: MFCD00077706
CAS Number: 22144-77-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty