Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Recombinant Streptavidin, Thermo Scientific

Supplier: Thermo Scientific
21122 21324 21122 21124
PI21122EA 235.2 USD InStock
PI21122 CAPI21122 CAPI21124 CAPI21324
Recombinant Streptavidin, Thermo Scientific
Proteins and Peptides
Use to develop in-house non-radioactive assay systems.

  • Sufficient For: Binding 17 to 24ug biotin per mg protein

Avidin can be used in ELISAs, immunohistochemical staining, Western blotting, hybridization, affinity chromatography, and fluorescent-activated cell sorting. The streptavidin conjugates are recommended for use where high binding affinity and specificity is required. NeutrAvidin® biotin-binding protein is a more economical alternative to both avidin and streptavidin. It features low nonspecific binding properties with no glycosylation. It also offers a neutral pI and lysine residues that remain available for derivatization or conjugation.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty