Would you like to log in again?
KIMAX® Short Cone Oil Centrifuge Tube, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
45243-100
89002-230CS 1422.63 USD InStock
89002-230
KIMAX® Short Cone Oil Centrifuge Tube, Kimble Chase
Tubes Centrifuge Tubes
Designed for field use in petroleum testing, these KIMAX® tubes are calibrated to contain and tooled to accept a snap cap.
  • Dimensions: 45Ext.Ø×165L mm (1³/₄×6").
  • Centrifuge tubes are made of borosilicate glass
Scale and legend are durable white ceramic enamel.
Certifications: Referenced in ASTM D4007.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty