Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc sulfate monohydrate USP

Supplier: RPI
Danger

Z22350-100.0
77510-516EA 13.5 USD InStock
77510-516
Zinc sulfate monohydrate USP
Zinc sulphate monohydrate
Caution: Research or further manufacturing use only, not for food or drug use.


Formula: ZnSO₄·1H₂O
MW: 179.45 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00149893
CAS Number: 7446-19-7
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance White Powder
Odor odorless
Solubility Soluble in Water and Glycerin. Insoluble in Alcohol
Identification (Zinc and Sulfate=+VE) Pass
Assay 99.0 - 100.5%
Acidity As per USP
Alkalies and Alkaline Earths ≤0.9%
Arsenic ≤0.0014%
Lead (Pb) ≤0.002%
Residual Solvents As per USP
Grade USP

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty